Learn More
ABclonal Technology Sin3A Rabbit pAb
SDP
Supplier:  ABclonal Technology A13928500UL
Encompass_Preferred
The protein encoded by this gene is a transcriptional regulatory protein. It contains paired amphipathic helix (PAH) domains, which are important for protein-protein interactions and may mediate repression by the Mad-Max complex.
Catalog No. 50-001-09000