Learn More
Aviva Systems Biology Corp SIN3B Antibody - N-terminal region (ARP36976_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36976_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SIN3B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2810430C10RikApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-56300