Learn More
Aviva Systems Biology Corp Sin3b Antibody - N-terminal region (ARP36977_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36977_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Sin3b. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse Sin3b. Alternate names: 2810430C10RikApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-014-56301