Learn More
Aviva Systems Biology Corp SIN3B Antibody - middle region (ARP50589_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50589_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIN3B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SIN3B. Alternate names: KIAA0700ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60382