Learn More
Cayman Chemical SInapaldehyde 50mg
SDP
Catalog No. 502943935
Encompass_Preferred
A phenylpropanoid with diverse biological activities; an inhibitor of FTase (IC50 = 23.99 µM); inhibits TNF-α-induced NF-κB transcriptional activity in a reporter assay using HepG2 cells (IC50 = 13.25 µM); reduces LPS-induced NO production in RAW 264.7 cells (IC50 = 5.92 µM); scavenges DPPH and ABTS radicals in cell-free assays (IC50s = 83.02 and 43.15 µM, respectively); active against Gram-positive and Gram-negative bacteria (MICs = 64-128 UG/ml); inhibits α-glucosidase (IC50 = 98.81 µM)
Catalog No. 50-294-3935 Supplier Cayman Chemical Supplier No. 4026750 MG
A phenylpropanoid with diverse biological activities; an inhibitor of FTase (IC50 = 23.99 µM); inhibits TNF-α-induced NF-κB transcriptional activity in a reporter assay using HepG2 cells (IC50 = 13.25 µM); reduces LPS-induced NO production in RAW 264.7 cells (IC50 = 5.92 µM); scavenges DPPH and ABTS radicals in cell-free assays (IC50s = 83.02 and 43.15 µM, respectively); active against Gram-positive and Gram-negative bacteria (MICs = 64-128 UG/ml); inhibits α-glucosidase (IC50 = 98.81 µM)