Learn More
Cayman Chemical Sinapic Acid, Purity: ≥98%, CAS: 530-59-6, 10 g
SDP
Catalog No. 502936259
Encompass_Preferred
A phenylpropanoid hydroxycinnamic acid with diverse biological activities; inhibits collagen-induced human platelet aggregation (IC50 = 1.03 mM); scavenges DPPH and ABTS free radicals (IC50s = 8.3 and 5.4 μg/ml, respectively); reduces colony formation of SW480 human colon carcinoma cells by 4-fold at 200 μM; inhibits colony formation of E. coli, S. enteritidis, and S. aureus on agar (MICs = 2.2, 2, and 1.8 mM, respectively); increases the time spent in the open arms of the elevated plus maze in mice at 4 mg/kg; commonly used as a matrix in protein mass spectrometry
Catalog No. 50-293-6259 Supplier Cayman Chemical Supplier No. 2545710 G
A phenylpropanoid hydroxycinnamic acid with diverse biological activities; inhibits collagen-induced human platelet aggregation (IC50 = 1.03 mM); scavenges DPPH and ABTS free radicals (IC50s = 8.3 and 5.4 μg/ml, respectively); reduces colony formation of SW480 human colon carcinoma cells by 4-fold at 200 μM; inhibits colony formation of E. coli, S. enteritidis, and S. aureus on agar (MICs = 2.2, 2, and 1.8 mM, respectively); increases the time spent in the open arms of the elevated plus maze in mice at 4 mg/kg; commonly used as a matrix in protein mass spectrometry