Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sinapic acid, Matrix Substance for MALDI-MS, ≥99.5%, MilliporeSigma™ Supelco™

Catalog No. 111014481 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 x 10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4481 10 x 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4481 Supplier MilliporeSigma Supelco Supplier No. 4950810X10MGF
Add to Cart
Add to Cart

Sinapic acid is one of the 4 most common hydroxycinnamic acids. It is widespread in the plant kingdom (vegetables, fruits, oilseed crops, cereal grains and medicinal plants, and some spices). It is mostly found in the form of esters, but may exist in the free form too.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 530-59-6
Linear Formula C11H12O5
MDL Number MFCD00004401
Melting Point 202°C
Synonym 3,5-Dimethoxy-4-hydroxycinnamic acid; 4-Hydroxy-3,5-dimethoxy-cinnamic acid; Sinapinic acid
Solubility Soluble in dioxane
Type Matrix Substance for MALDI-MS
Quantity 10 x 10 mg
Formula Weight 224.21
Percent Purity ≥99.5% (HPLC); ≥99.5%
Grade Matrix Substance for MALDI-MS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.