Learn More
Medchemexpress LLC Sinapine thiocyanat 250mg
SDP
Catalog No. 5000669459
Encompass_Preferred
Sinapine thiocyanate is an alkaloid isolated from seeds of the cruciferous species Sinapine thiocyanate exhibits anti-inflammatory anti-oxidant anti-tumor anti-angiogenic and radio-protective effects Sinapine thiocyanate is also an acetylcholinesterase (AChE) inhibitor and can be used for the research of Alzheimer s disease ataxia myasthenia gravis and Parkinson s disease[1][2][3][4]
Catalog No. 50-006-69459 Supplier Medchemexpress LLC Supplier No. 250MGHYN0450
Sinapine thiocyanate is an alkaloid isolated from seeds of the cruciferous species Sinapine thiocyanate exhibits anti-inflammatory anti-oxidant anti-tumor anti-angiogenic and radio-protective effects Sinapine thiocyanate is also an acetylcholinesterase (AChE) inhibitor and can be used for the research of Alzheimer s disease ataxia myasthenia gravis and Parkinson s disease[1][2][3][4]