Learn More
TARGETMOL CHEMICALS INC Sinapine thiocyanate 50MG
SDP
Catalog No. 502879499
Encompass_Preferred
Also available in 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Sinapine Thiocyanate, is a derivative of Sinapine (S486605), an alkaloidal amine found in black mustard seeds. It is the choline ester of Sinapic Acid (S486800). It is also a phenolic-chloine conjugates, acting as anti-oxidants and acetylcholinesterase inhibitors, that can be used for the treatment of Alzheimer's disease (AD). Purity 98%
Catalog No. 50-287-9499 Supplier TARGETMOL CHEMICALS INC Supplier No. T339250MG
Also available in 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Sinapine Thiocyanate, is a derivative of Sinapine (S486605), an alkaloidal amine found in black mustard seeds. It is the choline ester of Sinapic Acid (S486800). It is also a phenolic-chloine conjugates, acting as anti-oxidants and acetylcholinesterase inhibitors, that can be used for the treatment of Alzheimer's disease (AD). Purity 98%