Learn More
Medchemexpress LLC Sinapine thiocyanate | 7431-77-8 | 368.45 | 100 MG
SDP
Catalog No. 5000389688
Encompass_Preferred
Sinapine thiocyanate is an alkaloid isolated from seeds of cruciferous species. It exhibits anti-inflammatory, anti-oxidant, anti-tumor, anti-angiogenic, and radio-protective effects. This compound is also an acetylcholinesterase (AChE) inhibitor and can be used for the research of Alzheimer's disease, ataxia, myasthenia gravis, and Parkinson's disease.
  • Alkaloid isolated from seeds of cruciferous species.
  • Exhibits anti-inflammatory effects.
  • Exhibits anti-oxidant effects.
  • Exhibits anti-tumor effects.
  • Exhibits anti-angiogenic effects.
  • Exhibits radio-protective effects.
  • Acetylcholinesterase (AChE) inhibitor.
  • Used for research of Alzheimer's disease, ataxia, myasthenia gravis, and Parkinson's disease.
Catalog No. 50-003-89688 Supplier Medchemexpress LLC Supplier No. HYN0450100MG
Sinapine thiocyanate is an alkaloid isolated from seeds of cruciferous species. It exhibits anti-inflammatory, anti-oxidant, anti-tumor, anti-angiogenic, and radio-protective effects. This compound is also an acetylcholinesterase (AChE) inhibitor and can be used for the research of Alzheimer's disease, ataxia, myasthenia gravis, and Parkinson's disease.
  • Alkaloid isolated from seeds of cruciferous species.
  • Exhibits anti-inflammatory effects.
  • Exhibits anti-oxidant effects.
  • Exhibits anti-tumor effects.
  • Exhibits anti-angiogenic effects.
  • Exhibits radio-protective effects.
  • Acetylcholinesterase (AChE) inhibitor.
  • Used for research of Alzheimer's disease, ataxia, myasthenia gravis, and Parkinson's disease.