Learn More
Medchemexpress LLC Sinapine thiocyanate | 7431-77-8 | 368.45 | 50 MG
SDP
Catalog No. 5000390102
Encompass_Preferred
Sinapine thiocyanate is an alkaloid isolated from seeds of cruciferous species. It demonstrates anti-inflammatory, anti-oxidant, anti-tumor, anti-angiogenic, and radio-protective effects. It is also an acetylcholinesterase (AChE) inhibitor. It can be used for the research of Alzheimer's disease, ataxia, myasthenia gravis, and Parkinson's disease.
  • Exhibits anti-inflammatory effects.
  • Acts as an anti-oxidant.
  • Demonstrates anti-tumor and anti-angiogenic properties.
  • Provides radio-protective effects.
  • Inhibits acetylcholinesterase (AChE).
  • Suitable for research on neurological conditions like Alzheimer's and Parkinson's disease.
Catalog No. 50-003-90102 Supplier Medchemexpress LLC Supplier No. HYN045050MG
Sinapine thiocyanate is an alkaloid isolated from seeds of cruciferous species. It demonstrates anti-inflammatory, anti-oxidant, anti-tumor, anti-angiogenic, and radio-protective effects. It is also an acetylcholinesterase (AChE) inhibitor. It can be used for the research of Alzheimer's disease, ataxia, myasthenia gravis, and Parkinson's disease.
  • Exhibits anti-inflammatory effects.
  • Acts as an anti-oxidant.
  • Demonstrates anti-tumor and anti-angiogenic properties.
  • Provides radio-protective effects.
  • Inhibits acetylcholinesterase (AChE).
  • Suitable for research on neurological conditions like Alzheimer's and Parkinson's disease.