Learn More
Medchemexpress LLC Sinapyl alcohol-d3 5mg
SDP
Catalog No. 5000205684
Encompass_Preferred
Sinapyl alcohol-d3 is the deuterated labeled Sinapyl alcohol (HY-W109812) Sinapyl alcohol is an orally active anti-inflammatory and antinociceptive agent Sinapyl alcohol reduces the expression level of inducible NO synthase and COX-2[1]
Catalog No. 50-002-05684 Supplier Medchemexpress LLC Supplier No. HYW109812S5MG
Sinapyl alcohol-d3 is the deuterated labeled Sinapyl alcohol (HY-W109812) Sinapyl alcohol is an orally active anti-inflammatory and antinociceptive agent Sinapyl alcohol reduces the expression level of inducible NO synthase and COX-2[1]