Learn More
Bioworld SINCALIDE
Supplier:  Bioworld 220606951
Encompass_Preferred
Sincalide Source: Description : Sincalide also called CCK-8 has a molecular formula of C49H62N10O16S3 H-Asp-Tyr(SO3H)-Met-Gly-Trp-Met-Asp-Phe-NH2 having a Mw of 1143.29. Sincalide corresponds to the C-terminal octapeptide of cholecys...
Catalog No. 50-253-795