Learn More
Medchemexpress LLC Sincalide | 25126-32-3 | MFCD00079849 | 98.7% | 1143.27 | 1 MG
SDP
Catalog No. 5000426357
Encompass_Preferred
Sincalide, also known as Cholecystokinin octapeptide (CCK-8), is a rapid-acting amino acid polypeptide hormone analogue of cholecystokinin (CCK). It is intended for intravenous use in postevacuation cholecystography and retains most of CCK's biological activities.
  • Promotes gallbladder contraction
  • Helps diagnose gallbladder and pancreas disorders
  • Increases bile secretion and relaxes the sphincter of Oddi
  • Inhibits myocardial fibrosis and reduces inflammation and apoptosis
  • Protects H9c2 cardiomyoblasts from Ang II-induced apoptosis via CCK1 receptor and PI3K/Akt pathway
  • Alleviates fibrosis and delays left ventricular remodeling in MI rat models
  • For research use only
Catalog No. 50-004-26357 Supplier Medchemexpress LLC Supplier No. HYP00931MG
Sincalide, also known as Cholecystokinin octapeptide (CCK-8), is a rapid-acting amino acid polypeptide hormone analogue of cholecystokinin (CCK). It is intended for intravenous use in postevacuation cholecystography and retains most of CCK's biological activities.
  • Promotes gallbladder contraction
  • Helps diagnose gallbladder and pancreas disorders
  • Increases bile secretion and relaxes the sphincter of Oddi
  • Inhibits myocardial fibrosis and reduces inflammation and apoptosis
  • Protects H9c2 cardiomyoblasts from Ang II-induced apoptosis via CCK1 receptor and PI3K/Akt pathway
  • Alleviates fibrosis and delays left ventricular remodeling in MI rat models
  • For research use only