Learn More
Medchemexpress LLC Sincalide (ammonium) | 70706-98-8 | 99.2% | 1160.30 | 1 MG
SDP
Catalog No. 5000366079
Encompass_Preferred
Sincalide ammonium is a rapid-acting amino acid polypeptide hormone analogue of cholecystokinin (CCK). This major bioactive segment of CCK is used intravenously in postevacuation cholecystography, retaining most of CCK's biological activities. It can promote gallbladder contraction, increase bile secretion, and relax the sphincter of Oddi, assisting in the diagnosis of gallbladder and pancreas disorders.
  • Rapid-acting amino acid polypeptide hormone analogue
  • Used for postevacuation cholecystography
  • Promotes gallbladder contraction, increases bile secretion, and relaxes sphincter of Oddi
  • Aids in diagnosis of gallbladder and pancreas disorders
  • Inhibits myocardial fibrosis, inflammation, apoptosis, and collagen deposition (in vitro)
  • Alleviates fibrosis and delays left ventricular remodeling in MI rat models (in vivo)
  • For research use only
Catalog No. 50-003-66079 Supplier Medchemexpress LLC Supplier No. 1MGHYP0093A
Add to Cart
Add to Cart
Sincalide ammonium is a rapid-acting amino acid polypeptide hormone analogue of cholecystokinin (CCK). This major bioactive segment of CCK is used intravenously in postevacuation cholecystography, retaining most of CCK's biological activities. It can promote gallbladder contraction, increase bile secretion, and relax the sphincter of Oddi, assisting in the diagnosis of gallbladder and pancreas disorders.
  • Rapid-acting amino acid polypeptide hormone analogue
  • Used for postevacuation cholecystography
  • Promotes gallbladder contraction, increases bile secretion, and relaxes sphincter of Oddi
  • Aids in diagnosis of gallbladder and pancreas disorders
  • Inhibits myocardial fibrosis, inflammation, apoptosis, and collagen deposition (in vitro)
  • Alleviates fibrosis and delays left ventricular remodeling in MI rat models (in vivo)
  • For research use only