Learn More
Medchemexpress LLC Sincalide (ammonium) | 70706-98-8 | 99.2% | 1160.30 | 10 MG
SDP
Catalog No. 5000426147
Encompass_Preferred
Sincalide ammonium is a rapid-acting amino acid polypeptide hormone analogue of cholecystokinin (CCK) for intravenous use in postevacuation cholecystography. It is a major bioactive segment of CCK that retains most of its biological activities. CCK-8 can promote gallbladder contraction by injection and helps diagnose gallbladder and pancreas disorders, increasing bile secretion, causing gallbladder contraction, and relaxing the sphincter of Oddi for bile drainage into the duodenum.
  • Significant inhibitory effect on myocardial fibrosis in noninfarcted areas
  • Positive role in fighting inflammation, apoptosis, and collagen deposition
  • Protects H9c2 cardiomyoblasts from Ang II‐induced apoptosis
  • Alleviates fibrosis in noninfarcted regions
  • Delays left ventricular remodeling and the progress of heart failure in a MI rat model
Catalog No. 50-004-26147 Supplier Medchemexpress LLC Supplier No. HYP0093A10MG
Sincalide ammonium is a rapid-acting amino acid polypeptide hormone analogue of cholecystokinin (CCK) for intravenous use in postevacuation cholecystography. It is a major bioactive segment of CCK that retains most of its biological activities. CCK-8 can promote gallbladder contraction by injection and helps diagnose gallbladder and pancreas disorders, increasing bile secretion, causing gallbladder contraction, and relaxing the sphincter of Oddi for bile drainage into the duodenum.
  • Significant inhibitory effect on myocardial fibrosis in noninfarcted areas
  • Positive role in fighting inflammation, apoptosis, and collagen deposition
  • Protects H9c2 cardiomyoblasts from Ang II‐induced apoptosis
  • Alleviates fibrosis in noninfarcted regions
  • Delays left ventricular remodeling and the progress of heart failure in a MI rat model