Learn More
Medchemexpress LLC Sincalide (ammonium) | 70706-98-8 | 99.2% | 1160.30 | 5 MG
SDP
Catalog No. 5000426031
Encompass_Preferred
Sincalide ammonium is a rapid-acting amino acid polypeptide hormone analogue of cholecystokinin (CCK) used for intravenous applications like postevacuation cholecystography. It is a major bioactive segment of CCK, retaining most of its biological activities. It can promote gallbladder contraction and aids in diagnosing gallbladder and pancreas disorders.
  • Increases bile secretion
  • Causes gallbladder to contract and relax the sphincter of Oddi, facilitating bile drainage
  • Shows inhibitory effect on myocardial fibrosis
  • Plays a positive role in fighting inflammation, apoptosis, and collagen deposition
Catalog No. 50-004-26031 Supplier Medchemexpress LLC Supplier No. HYP0093A5MG
Sincalide ammonium is a rapid-acting amino acid polypeptide hormone analogue of cholecystokinin (CCK) used for intravenous applications like postevacuation cholecystography. It is a major bioactive segment of CCK, retaining most of its biological activities. It can promote gallbladder contraction and aids in diagnosing gallbladder and pancreas disorders.
  • Increases bile secretion
  • Causes gallbladder to contract and relax the sphincter of Oddi, facilitating bile drainage
  • Shows inhibitory effect on myocardial fibrosis
  • Plays a positive role in fighting inflammation, apoptosis, and collagen deposition