Learn More
Enzo Life Sciences Sinefungin, (1mg), CAS Number: 58944-73-3
SDP
Catalog No. NC9800521
Encompass_Preferred
Methyltransferase inhibitor. Strong competitive inhibitor of methyltransferases which uses S-adenosyl-1-methionine as the methyl group donor to yield methylated products such as 5-methylcytosine or N6-methyl adenosine on DNA and RNA. In addition, sinefungin is involved in a number of physiological processes. Alternative Name: 6,9-Diamino-1-(6-amino-9H-purin-9-yl)-1,5,6,7,8,9-hexadeoxy-D-glycero-alpha-L-talo-decafuranuronic acid, A-9145, Adenosylornithine. Formula: C15H23N7O5. MW: 381.4. CAS Number: 58944-73-3. MI: 14: 8544. Purity: ≥95% (HPLC). Appearance: White to yellow powder. Solubility: Soluble in water. Long Term Storage: -20°C
Catalog No. NC9800521 Supplier Enzo Life Sciences Supplier No. ALX380070M001
Add to Cart
Add to Cart
Methyltransferase inhibitor. Strong competitive inhibitor of methyltransferases which uses S-adenosyl-1-methionine as the methyl group donor to yield methylated products such as 5-methylcytosine or N6-methyl adenosine on DNA and RNA. In addition, sinefungin is involved in a number of physiological processes. Alternative Name: 6,9-Diamino-1-(6-amino-9H-purin-9-yl)-1,5,6,7,8,9-hexadeoxy-D-glycero-alpha-L-talo-decafuranuronic acid, A-9145, Adenosylornithine. Formula: C15H23N7O5. MW: 381.4. CAS Number: 58944-73-3. MI: 14: 8544. Purity: ≥95% (HPLC). Appearance: White to yellow powder. Solubility: Soluble in water. Long Term Storage: -20°C