Learn More
Medchemexpress LLC HY-101938 1mg Medchemexpress, Sinefungin CAS:58944-73-3 Purity:>98%
SDP
Catalog No. 501967928
Encompass_Preferred
Medchemexpress, HY-101938 1mg Sinefungin CAS:58944-73-3 Sinefungin is a potent inhibitor of virion mRNA(guanine-7-)-methyltransferase, mRNA(nucleoside-2'-)-methyltransferase, and viral multiplication. Sinefungin, a SET7/9 inhibitor, ameliorates renal fibrosis by inhibiting H3K4 methylation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7928 Supplier Medchemexpress LLC Supplier No. HY1019381MG
Medchemexpress, HY-101938 1mg Sinefungin CAS:58944-73-3 Sinefungin is a potent inhibitor of virion mRNA(guanine-7-)-methyltransferase, mRNA(nucleoside-2'-)-methyltransferase, and viral multiplication. Sinefungin, a SET7/9 inhibitor, ameliorates renal fibrosis by inhibiting H3K4 methylation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.