Learn More
Azzota Corp Single Weighting Technical Balance, 6500G Capacity, 0.1G Readability, +/-0.1G Linearity, Dia. 160mm Pan
Catalog No. 501613023
Encompass_Preferred
Single weighting technical Balance, 6500g Capacity, 0.1g Readability, +/-0.1g Linearity, Dia. 160mm Pan
Catalog No. 50-161-3023 Supplier Azzota Corp Supplier No. TSM6500
Single weighting technical Balance, 6500g Capacity, 0.1g Readability, +/-0.1g Linearity, Dia. 160mm Pan