Learn More
MSC Singer Safety Acoustic Foam, 0.85 NRC, 27x54x3in, 4/Pack
Catalog No. 5000795629
Encompass
This acoustic foam is made from polyester polyurethane, designed to reduce noise with a Noise Reduction Coefficient Rating (NRCR) of 0.85. It features a plain backing and surface, making it suitable for various sound dampening applications.
  • Made from polyester polyurethane
  • Features a 0.85 noise reduction coefficient rating
  • Plain backing for easy application
  • Complies with Dept of Transportation MVSS #302 and Underwriters Laboratories UL94 #HF-1 standards
  • Operates at temperatures as low as -40°F
  • Dimensions of 27 in long, 54 in wide, and 3 in thick
Catalog No. 50-007-95629 Supplier MSC Supplier No. 42129254
This acoustic foam is made from polyester polyurethane, designed to reduce noise with a Noise Reduction Coefficient Rating (NRCR) of 0.85. It features a plain backing and surface, making it suitable for various sound dampening applications.
  • Made from polyester polyurethane
  • Features a 0.85 noise reduction coefficient rating
  • Plain backing for easy application
  • Complies with Dept of Transportation MVSS #302 and Underwriters Laboratories UL94 #HF-1 standards
  • Operates at temperatures as low as -40°F
  • Dimensions of 27 in long, 54 in wide, and 3 in thick