Learn More
Analytical Sales & Services FlexChrom Single UHPLC Assembly – 30cm x 0.010" ID
Supplier:  Analytical Sales & Services 970128FLEX
Encompass
FlexChrom, Single 1/16 in. Flexible UHPLC Assembly, 30cm x 0.010 in. ID (Ends of Tubing are 1/16 in., Body is Ground down to 1/32 in. to Allow Flexibility)
Catalog No. 50-456-290