Learn More
World Precision Instrument Single Animal Vaporizer Unit
SDP
Catalog No. 502538855
Encompass
Vaporizer for precise blend of oxygen and isoflurane
The vaporizer is a precision instrument that blends incoming oxygen with liquid isoflurane and outputs anesthetic gas at the set oxygen to isoflurane ratio. The dial setting allows fine gradations for precise isoflurane concentration. The vaporizer is available in three configurations that incorporate output to an anesthesia system or as a stand-alone unit. The Classic Anesthesia System (EZ-7000) and the Single Animal Anesthesia System (EZ-B800)  are also available without a vaporizer, allowing you to incorporate your own unit.
Catalog No. 50-253-8855 Supplier World Precision Instrument Supplier No. EZ108SA
Vaporizer for precise blend of oxygen and isoflurane
The vaporizer is a precision instrument that blends incoming oxygen with liquid isoflurane and outputs anesthetic gas at the set oxygen to isoflurane ratio. The dial setting allows fine gradations for precise isoflurane concentration. The vaporizer is available in three configurations that incorporate output to an anesthesia system or as a stand-alone unit. The Classic Anesthesia System (EZ-7000) and the Single Animal Anesthesia System (EZ-B800)  are also available without a vaporizer, allowing you to incorporate your own unit.