Learn More
Accuflow Systems Single braid Reinforced Tubing
Supplier: Accuflow Systems ONESILSB05000875
Purity One™ OneSil single-braid reinforced biopharmaceutical grade silicone tubing has been formulated for demanding high-pressure applications in biopharmaceutical processing. This flexible silicone tubing is moldable for single-use assemblies and has higher pressure ratings than non-reinforced tubing, making it an excelling choice for bulk transfer, SIP/CIP, filtration, and buffer and media prep. Full validation guide and USP <665> Extractables testing available.