Learn More
Grainger Single Calibration Gas NO2 29L 10PPM
Catalog No. 502750144
Encompass_Preferred
The Single Calibration Gas NO2 29L 10PPM is a calibration gas cylinder containing 10 parts per million of nitrogen dioxide (NO2) in a balance of nitrogen. It has a volume of 29 liters and is designed for calibration and bump testing of gas detection instruments. The cylinder has a CGA-590 outlet connection and comes equipped with a carrying strap for portability. The gas mixture is certified and traceable to NIST standards.
Catalog No. 50-275-0144 Supplier Grainger Supplier No. 156A92
The Single Calibration Gas NO2 29L 10PPM is a calibration gas cylinder containing 10 parts per million of nitrogen dioxide (NO2) in a balance of nitrogen. It has a volume of 29 liters and is designed for calibration and bump testing of gas detection instruments. The cylinder has a CGA-590 outlet connection and comes equipped with a carrying strap for portability. The gas mixture is certified and traceable to NIST standards.