Learn More
Agilent Technologies Reservoir, Single Cavity, Irradiated Polypropylene, 300 mL, 25/pk
Catalog No. NC1011368
Encompass
Reservoir, single cavity, irradiated polypropylene, 300 mL, 96 pyramids base geometry, 44 mm height, individually bagged, 25/pk.
Catalog No. NC1011368 Supplier Agilent Technologies Supplier No. 204017100
Reservoir, single cavity, irradiated polypropylene, 300 mL, 96 pyramids base geometry, 44 mm height, individually bagged, 25/pk.