Learn More
Bellco Glass, Inc. Single Cavity Reagent
SDP
Catalog No. 502330528
Encompass_Preferred
DescriptionReagent Reservoirs are designed to assist in repetitive or robotic liquid transferPyramid design promotes funneling of liquid to maximize reagent recovery by pipet tipsLow profile is ideal for small volume robotic tipsPolypropylene offers a low binding surface to prevent samples from sticking to the side during elution, and is chemically inert for combinatorial chemistry applicationsAutoclavable Pack Description5/BagCase Quantity25SterileNoNon-pyrogenicYesAutoclavableYesPrimary ColorNaturalPrimary MaterialPolypropyleneMaximum Volume195mLRNase/DNase FreeYes
Catalog No. 50-233-0528 Supplier Bellco Glass, Inc. Supplier No. 229563000
DescriptionReagent Reservoirs are designed to assist in repetitive or robotic liquid transferPyramid design promotes funneling of liquid to maximize reagent recovery by pipet tipsLow profile is ideal for small volume robotic tipsPolypropylene offers a low binding surface to prevent samples from sticking to the side during elution, and is chemically inert for combinatorial chemistry applicationsAutoclavable Pack Description5/BagCase Quantity25SterileNoNon-pyrogenicYesAutoclavableYesPrimary ColorNaturalPrimary MaterialPolypropyleneMaximum Volume195mLRNase/DNase FreeYes