Learn More
Perkin Elmer US LLC Single Cell HEN MicroFlow Nebulizer for NexION 300/350
Catalog No. 502112037
Encompass_Preferred
When combined with the Asperon Spray Chamber (N8152493), the HEN Nebulizer delivers cells intact to the ionization source. It is included in the Single Cell Sample Introduction Kit for NexION 300/350 (N8140032).
Catalog No. 50-211-2037 Supplier Perkin Elmer US LLC Supplier No. N8142046
Add to Cart
Add to Cart
Request a Quote
When combined with the Asperon Spray Chamber (N8152493), the HEN Nebulizer delivers cells intact to the ionization source. It is included in the Single Cell Sample Introduction Kit for NexION 300/350 (N8140032).