Learn More
Perkin Elmer US LLC Single Cell HEN MicroFlow Nebulizer for NexION 1000/2000
Catalog No. 502112949
Encompass_Preferred
When combined with the Asperon Spray Chamber (N8152493), the HEN Nebulizer delivers cells intact to the ionization source. It is included in the Single Cell Sample Introduction Kit for NexION 1000/2000 (N8150032).
Catalog No. 50-211-2949 Supplier Perkin Elmer US LLC Supplier No. N8152496
When combined with the Asperon Spray Chamber (N8152493), the HEN Nebulizer delivers cells intact to the ionization source. It is included in the Single Cell Sample Introduction Kit for NexION 1000/2000 (N8150032).