Learn More
Perkin Elmer US LLC Single Cell Holder for LAMBDA 365 - 10 mm
Catalog No. 502105529
Encompass_Preferred
This Single Cell Holder accommodates the double-beam configuration of the LAMBDA 365. It holds two standard, rectangular 10 mm cells (27.5 x 12.5 x 12.5 mm): one for the sample being tested, the other for a reference sample.
Catalog No. 50-210-5529 Supplier Perkin Elmer US LLC Supplier No. N4101027
This Single Cell Holder accommodates the double-beam configuration of the LAMBDA 365. It holds two standard, rectangular 10 mm cells (27.5 x 12.5 x 12.5 mm): one for the sample being tested, the other for a reference sample.