Learn More
Perkin Elmer US LLC Single Cell Holder for LAMBDA 465 - 10 mm
Catalog No. 502105477
Encompass_Preferred
This Single Cell Holder positions samples so they take full advantage of the the LAMBDA 465's dual - deuterium and tungsten - light source, ensuring precise sample alignment.
Catalog No. 50-210-5477 Supplier Perkin Elmer US LLC Supplier No. N4104000
This Single Cell Holder positions samples so they take full advantage of the the LAMBDA 465's dual - deuterium and tungsten - light source, ensuring precise sample alignment.