Learn More
Perkin Elmer US LLC Single Cell Peltier Accessory for Fluorescence
Catalog No. 502112754
Encompass_Preferred
This accessory is the ideal choice for a wide variety of applications such as the thermal denaturation of proteins. It is compatible with the LL 45/50/55 fluorescence spectrometers. It requires the PCB Accessory Kit for use with the LS 45.
Catalog No. 50-211-2754 Supplier Perkin Elmer US LLC Supplier No. L2250150
Add to Cart
Add to Cart
Request a Quote
This accessory is the ideal choice for a wide variety of applications such as the thermal denaturation of proteins. It is compatible with the LL 45/50/55 fluorescence spectrometers. It requires the PCB Accessory Kit for use with the LS 45.