Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.
Specifications
Specifications
| Description | Long path cylindrical cell holder |
| Type | Long Path Cell Holder |
| For Use With (Application) | Measurements of gases and select older standard methods that call specifically for cylindrical cells |
| For Use With (Equipment) | GENESYS 140/150/180,BioMate160 |
| Holds | 5 to 100mm cylindrical cuvette |
| Shape | Cylindrical |