Learn More
Perkin Elmer US LLC Single Cell Sample Introduction Kit for NexION 1000/2000
Supplier:  Perkin Elmer US LLC N8150032
Encompass_Preferred
The Single Cell ICP-MS Sample Introduction Kits are part of PerkinElmers award-winning NexION ICP-MS Single Cell Analysis System. They include our proprietary Asperon Spray Chamber and HEN Single Cell Nebulizer as well as all of the additional consumables needed for single cell analysis with your NexION ICP-MS system.
Catalog No. 50-210-4696
Add to Cart
Request a Quote