Learn More
Waters Corp Waters Single-channel Pipette, 50-1000 μL, Electronic, Ergonomic
Catalog No. 502269327 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Bluetooth electronic single-channel pipettes deliver accurate and precise liquid handling. Featuring an ergonomic and lightweight design, they help prevent repetitive strain injuries by promoting an optimal working posture. Fully electronic operation minimizes user variability, and Bluetooth connectivity enables remote programming and pipetting traceability for enhanced workflow management.
  • Bluetooth capability for two-way communication, remote programming, and pipetting traceability.
  • Intuitive user interface.
  • Autoclavable lower parts (excluding 1200 μL multi-channel lower parts).
  • Optiload for perfect tip sealing.
  • Optimized pipetting modes for multi-dispensing, multi-aspiration, reverse pipetting, mixing, and more.
  • Safe-cone filters in all models >10 μL to prevent contamination.
This item is not returnable. View return policy
Catalog No. 50-226-9327 Supplier Waters Corp Supplier No. 186009766
These Bluetooth electronic single-channel pipettes deliver accurate and precise liquid handling. Featuring an ergonomic and lightweight design, they help prevent repetitive strain injuries by promoting an optimal working posture. Fully electronic operation minimizes user variability, and Bluetooth connectivity enables remote programming and pipetting traceability for enhanced workflow management.
  • Bluetooth capability for two-way communication, remote programming, and pipetting traceability.
  • Intuitive user interface.
  • Autoclavable lower parts (excluding 1200 μL multi-channel lower parts).
  • Optiload for perfect tip sealing.
  • Optimized pipetting modes for multi-dispensing, multi-aspiration, reverse pipetting, mixing, and more.
  • Safe-cone filters in all models >10 μL to prevent contamination.