Learn More
Waters Corp Andrew Alliance Pipette, 8-channel, 5-120 μL
Catalog No. 502269321 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Andrew Alliance Bluetooth Electronic 8-channel Pipettes provide accurate and precise pipetting for use with Andrew+ pipetting robot, Pipette+ guided system, or manual use. Based on the Sartorius Picus Pipette, the ergonomic and lightweight design prevents repetitive strain injuries. Fully electronic operation reduces user variability. Bluetooth enables two-way communication with Andrew+ or Pipette+ and OneLab for remote programming and traceability.
  • Intuitive user interface
  • Autoclavable lower parts (except for 1200 μL multi-channel lower parts)
  • Optiload for perfect tip sealing
  • Optimized pipetting modes for multi-dispensing, multi-aspiration, reverse pipetting, mixing, and more
  • Safe-cone filters in all models >10 μL to prevent contamination
This item is not returnable. View return policy
Catalog No. 50-226-9321 Supplier Waters Corp Supplier No. 186009605
Add to Cart
Add to Cart
These Andrew Alliance Bluetooth Electronic 8-channel Pipettes provide accurate and precise pipetting for use with Andrew+ pipetting robot, Pipette+ guided system, or manual use. Based on the Sartorius Picus Pipette, the ergonomic and lightweight design prevents repetitive strain injuries. Fully electronic operation reduces user variability. Bluetooth enables two-way communication with Andrew+ or Pipette+ and OneLab for remote programming and traceability.
  • Intuitive user interface
  • Autoclavable lower parts (except for 1200 μL multi-channel lower parts)
  • Optiload for perfect tip sealing
  • Optimized pipetting modes for multi-dispensing, multi-aspiration, reverse pipetting, mixing, and more
  • Safe-cone filters in all models >10 μL to prevent contamination