Learn More
Waters Corp Andrew Alliance Pipette, 8-channel, 0.2-10 μL, Bluetooth Electronic
Catalog No. 502269329 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Andrew Alliance Bluetooth Electronic 8-channel Pipettes offer accurate and precise pipetting. They are designed for use with the Andrew+ pipetting robot, the Pipette+ guided pipetting system, or for manual pipetting. The pipette is based on the Sartorius Picus Pipette and features an ergonomic and lightweight design to prevent repetitive strain injuries by promoting an optimal working posture and comfortable pipetting. Fully electronic operation helps reduce variability between users. The Bluetooth capability allows for two-way communication when used with Andrew+ or Pipette+ and OneLab, enabling remote programming of pipettes and providing pipetting traceability.
  • Intuitive user interface
  • Autoclavable lower parts (except for 1200 μL multi-channel lower parts)
  • Optiload for perfect tip sealing
  • Optimized pipetting modes for multi-dispensing, multi-aspiration, reverse pipetting, mixing and more
This item is not returnable. View return policy
Catalog No. 50-226-9329 Supplier Waters Corp Supplier No. 186009768
Add to Cart
Add to Cart
Andrew Alliance Bluetooth Electronic 8-channel Pipettes offer accurate and precise pipetting. They are designed for use with the Andrew+ pipetting robot, the Pipette+ guided pipetting system, or for manual pipetting. The pipette is based on the Sartorius Picus Pipette and features an ergonomic and lightweight design to prevent repetitive strain injuries by promoting an optimal working posture and comfortable pipetting. Fully electronic operation helps reduce variability between users. The Bluetooth capability allows for two-way communication when used with Andrew+ or Pipette+ and OneLab, enabling remote programming of pipettes and providing pipetting traceability.
  • Intuitive user interface
  • Autoclavable lower parts (except for 1200 μL multi-channel lower parts)
  • Optiload for perfect tip sealing
  • Optimized pipetting modes for multi-dispensing, multi-aspiration, reverse pipetting, mixing and more