Learn More
Waters Corp Andrew Alliance Pipette, Single-channel, 5–120 μL
Catalog No. 502269326 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Bluetooth Electronic Single-channel Pipettes offer accurate and precise liquid handling. Designed for use with Andrew+ robots, Pipette+ systems, or manual operation, they are based on the Sartorius Picus Pipette. Their ergonomic design helps prevent repetitive strain injuries. Fully electronic operation minimizes variability, while Bluetooth enables remote programming and traceability with compatible systems like Andrew+ or Pipette+ and OneLab.
  • Intuitive user interface
  • Autoclavable lower parts (except for 1200 μL multi-channel lower parts)
  • Optiload for perfect tip sealing
  • Optimized pipetting modes for multi-dispensing, multi-aspiration, reverse pipetting, mixing and more
  • Safe-cone filters in all models >10 μL to prevent contamination
This item is not returnable. View return policy
Catalog No. 50-226-9326 Supplier Waters Corp Supplier No. 186009765
These Bluetooth Electronic Single-channel Pipettes offer accurate and precise liquid handling. Designed for use with Andrew+ robots, Pipette+ systems, or manual operation, they are based on the Sartorius Picus Pipette. Their ergonomic design helps prevent repetitive strain injuries. Fully electronic operation minimizes variability, while Bluetooth enables remote programming and traceability with compatible systems like Andrew+ or Pipette+ and OneLab.
  • Intuitive user interface
  • Autoclavable lower parts (except for 1200 μL multi-channel lower parts)
  • Optiload for perfect tip sealing
  • Optimized pipetting modes for multi-dispensing, multi-aspiration, reverse pipetting, mixing and more
  • Safe-cone filters in all models >10 μL to prevent contamination