Learn More
Integra Biosciences Corp SINGLE CHANNEL EVOLVE STARTER
SDP
Catalog No. NC1835297
Encompass
Single Channel EVOLVE Starter PackTraditional pipettes utilize a single rotating plunger to set the volumes. This approach makes frequent adjustment of volumes tedious, especially when switching from lower to higher volumes.To overcome this pain point, the EVOLVE manual pipette has been designed with three adjustable dials to rapidly set volumes. This revolutionary approach reduces repetitive stress injuries and allows users to set volumes in a fraction of the time it takes with a traditional manual pipette.Includes 20, 200 and 1000 l single channel EVOLVE single channel pipettes.Includes shelf hook and one rack of each volume GripTip. GripTips snap into place and never loosen, leak or fall off.
Catalog No. NC1835297 Supplier Integra Biosciences Corp Supplier No. 3050
Single Channel EVOLVE Starter PackTraditional pipettes utilize a single rotating plunger to set the volumes. This approach makes frequent adjustment of volumes tedious, especially when switching from lower to higher volumes.To overcome this pain point, the EVOLVE manual pipette has been designed with three adjustable dials to rapidly set volumes. This revolutionary approach reduces repetitive stress injuries and allows users to set volumes in a fraction of the time it takes with a traditional manual pipette.Includes 20, 200 and 1000 l single channel EVOLVE single channel pipettes.Includes shelf hook and one rack of each volume GripTip. GripTips snap into place and never loosen, leak or fall off.