Learn More
Checkers Single Channel Rubber Duct Cable Protector
Catalog No. 19088433 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
19-088-433 Black
1 options
Catalog No. 19-088-433 Supplier Checkers Supplier No. RFD75
Only null left
Add to Cart
Add to Cart
Rubber duct systems protect and hide valuable cords and cables located in walkways or work areas. These non-metallic, flexible, industrial-grade rubber protectors are extremely durable, easy to install and organize portable cords and cables while reducing trip hazards.
  • Single Channel Protector For A Phone Line Or Small 14 Through 22 Gauge Wire
  • Can Be Slit If Necessary (Supplied Unslit)
  • Reduces Tripping Hazards
  • Protects And Organizes Portable Cords And Cables
  • Non-Metallic, Industrial-Grade Rubber Construction
  • Multiple Sizes For Various Cable Diameters
  • Grooved Underside For Better Traction
  • Optional Connector Kit Available For Most Sizes
Specifications
Material Rubber
Color Black
Product Type Cable Protector