Learn More
Cole-Parmer SINGLE CHANNEL TRACEABLE TIMER
Supplier:  Cole-Parmer 9446115
Encompass_Preferred
Traceable Flashing LED Big-Digit Timer with Calibration; Single-Channel.Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratoryStore three countdown times in memory for your most common usesTraceable model number 5013
Catalog No. NC2215639