Learn More
Jorgensen Labs Single Collection Unit Prefilled CPDA-1, 250mL
SDP
Catalog No. NC1665211
Encompass_Preferred
Single Collection Unit Prefilled CPDA-1. CPDA-1 is the anticoagulant of choice in human blood banking. After collection, stored blood will have a 35-day shelf life. Bag comes with 16g diamond point non-detachable needle plus double spike ports. Single Collection Unit Prefilled CPDA-1. 250ml. Contains 35ml of CPDA-1. For veterinary use only
This item is not returnable. View return policy
Catalog No. NC1665211 Supplier Jorgensen Labs Supplier No. J0520Q
Single Collection Unit Prefilled CPDA-1. CPDA-1 is the anticoagulant of choice in human blood banking. After collection, stored blood will have a 35-day shelf life. Bag comes with 16g diamond point non-detachable needle plus double spike ports. Single Collection Unit Prefilled CPDA-1. 250ml. Contains 35ml of CPDA-1. For veterinary use only