Learn More
Electron Microscopy Sciences Single Cross Line Reticle RET 43, 21 mm, 25 µm Line width
SDP
Catalog No. 501901744
Encompass_Preferred
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 21 mm
Line width: 25 µm
Catalog No. 50-190-1744 Supplier Electron Microscopy Sciences Supplier No. 6025221
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 21 mm
Line width: 25 µm