Learn More
Innovative Research Inc Single Donor Human Plasma (Blood Derived) K2 EDTA 10ml
Catalog No. 502036348
Encompass_Preferred
Single Donor Human Plasma from Innovative Research is whole blood derived. The whole blood is collected from donors in an FDA-approved collection center and processed into plasma via cetrifugation. The plasma is frozen immediately after processing.
Viral Testing
Each unit is tested for the standard FDA-required viral markers, and found negative for HBsAg, HCV, HIV-1, HIV-2, HIV-1Ag or HIV-1 NAT, ALT, and syphilis using FDA-approved methods. If you would like any further testing or analysis performed prior to shipping, please let us know. 
Donor Specifications
Innovative Research has a wide network of FDA-approved collection centers across the United States, which allows us to collect from nearly any donor type. We are able to accommodate demographic requests from a variety of genders, races, and ages. We are also able to work with custom collection requests and protocols. If you have custom demographic or collection requests, please let us know.
Catalog No. 50-203-6348 Supplier Innovative Research Inc Supplier No. IPLASK2E10ML
Single Donor Human Plasma from Innovative Research is whole blood derived. The whole blood is collected from donors in an FDA-approved collection center and processed into plasma via cetrifugation. The plasma is frozen immediately after processing.
Viral Testing
Each unit is tested for the standard FDA-required viral markers, and found negative for HBsAg, HCV, HIV-1, HIV-2, HIV-1Ag or HIV-1 NAT, ALT, and syphilis using FDA-approved methods. If you would like any further testing or analysis performed prior to shipping, please let us know. 
Donor Specifications
Innovative Research has a wide network of FDA-approved collection centers across the United States, which allows us to collect from nearly any donor type. We are able to accommodate demographic requests from a variety of genders, races, and ages. We are also able to work with custom collection requests and protocols. If you have custom demographic or collection requests, please let us know.