Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOMNI Scientific™ Single E-cylinder Manifold
SDP

Catalog No. 04777125 Shop All Somni Scientific Products
Change view
Click to view available options
Length (Metric):
14.6 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
04-777-125 14.6 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-777-125 Supplier SOMNI Scientific™ Supplier No. RE20101
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Can be used with a single E size oxygen cylinder or as part of an ancillary system with an oxygen concentrator

  • Attach one size E cylinder
  • Ancillary option with an oxygen concentrator 
  • Preset regulator delivers 50-55 psi
  • Durable, compact and easy to install
  • Preset regulator, mounting hardware, oxygen hose and cylinder wrench included

Specifications

For Use With (Application) Animal Oxygen Supply, Inhalant Anesthesia
For Use With (Equipment) RM-17003, RM-17004, OC-21003, OC-21004, MW-17005
Material Stainless Steel, Aluminum
Length (Metric) 14.6 cm
Width (Metric) 15.2 cm
Includes Preset regulator, mounting hardware, oxygen hose and cylinder wrench

For Veterinary Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.