Learn More
RAZOR BLADE COMPANY SINGLE EDGE BLADES KEEN EDGE
Supplier:  RAZOR BLADE COMPANY 940474
Encompass_Preferred
Unbacked, Uncoated, Degreased single edge blades stainless steel-x-keen edge-.009" Thisck 100 blades per carton/10 carton per case
Catalog No. NC1284376