Learn More
Electron Microscopy Sciences Razor Blade Single Edge Gem
SDP
Catalog No. 50949322
Encompass_Preferred
Carbon steel, .009" thick, 1 1/2"x 3/4" size, packed 10 blades in a dispenser.
Catalog No. 50-949-322 Supplier Electron Microscopy Sciences Supplier No. 71951
Carbon steel, .009" thick, 1 1/2"x 3/4" size, packed 10 blades in a dispenser.