Learn More
Electron Microscopy Sciences Razor Blade Single Edge GEM, Washed, Clamshell, PTFE Coated
SDP
Supplier:  Electron Microscopy Sciences 71970WA
Encompass_Preferred
GEM "PTFE" Coated and uncoated extra keen are made from stainless steel. easures 0.009" thick and 1 1/2"x¾" size. Washed and degreased, individually wrapped in a box to reduce the amount of foreign material on the blade. Clean room classified.
Coating: PTFE Coated
Catalog No. NC0991551