Learn More
Cpi International SINGLE ELEMENT CAL
SDP
Catalog No. NC9565451
Encompass
CPI International's Peak Performance Calcium (Ca) standard is provided at a concentration of 10,000 ppm and a volume of 100 mL. 10,000 ug/mL. In 4% HNO3 Matrix - 18 month shelf-life.
Catalog No. NC9565451 Supplier Cpi International Supplier No. S440010M91
CPI International's Peak Performance Calcium (Ca) standard is provided at a concentration of 10,000 ppm and a volume of 100 mL. 10,000 ug/mL. In 4% HNO3 Matrix - 18 month shelf-life.