Learn More
Cpi International SINGLE ELEMENT STANDARD ALUMI
SDP
Catalog No. NC9117712
Encompass
Single-Element Standards 1000microgram/mL, Element: Aluminum, Source: Al metal, Purity: 99.999%, Matrix: 2% HNO3
Catalog No. NC9117712 Supplier Cpi International Supplier No. S4400100011
Add to Cart
Add to Cart
Single-Element Standards 1000microgram/mL, Element: Aluminum, Source: Al metal, Purity: 99.999%, Matrix: 2% HNO3